Methyl-5-amino-2-methylbenzoate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M2540-25G M2540-5G
TCM2540-25GEA 486.64 CAD
TCM2540-25G TCM2540-5G
Methyl-5-amino-2-methylbenzoate ≥98.0% (by GC, titration analysis)
Methyl-5-amino-2-methylbenzoate

CAS Number: 18595-12-5
MDL Number: MFCD08752568
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 42
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 5-Amino-2-methylbenzoic Acid Methyl Ester, Methyl 5-Amino-o-toluate, 5-Amino-o-toluic Acid Methyl Ester

Formula: C₉H₁₁NO₂
Melting Pt: 42 °C
MDL Number: MFCD08752568
CAS Number: 18595-12-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a24fe9e6c48692c8b14109d9280dd094d8a9390&vid=4a24fe9e6c48692c8b14109d9280dd094d8a9390&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR