Methyl-4-fluoro-3-nitrobenzoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 4-fluoro-3-nitrobenzoate, 4-Fluorobenzoic acid methyl ester

M2535-1G M2535-5G
TCM2535-1GEA 50.56 CAD
TCM2535-1G TCM2535-5G
Methyl-4-fluoro-3-nitrobenzoate ≥98.0% (by GC)
Methyl 4-fluoro-3-nitrobenzoate

CAS Number: 329-59-9
MDL Number: MFCD00196161
Molecular Formula: C8H6FNO4
Molecular Weight: 199.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 119
Melting point (°C): 61
Flash Point (°C): 110


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoro-3-nitrobenzoic Acid Methyl Ester

Formula: C₈H₆FNO₄
MW: 199.14 g/mol
Boiling Pt: 119 °C
Melting Pt: 61 °C
Flash Pt: 110 °C
MDL Number: MFCD00196161
CAS Number: 329-59-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=683a58da1091aa6157a9567def1e7a1d05f7fc87&vid=683a58da1091aa6157a9567def1e7a1d05f7fc87&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR