Maprotiline hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 9-(gamma-Methylaminopropyl)-9,10-dihydro-9,10-ethanoanthracene Hydrochloride

M2527-1G M2527-5G
TCM2527-1GEA 112.34 CAD
TCM2527-1G TCM2527-5G
Maprotiline hydrochloride ≥98.0%
Maprotiline hydrochloride

CAS Number: 10347-81-6
MDL Number: MFCD00079464
Molecular Formula: C20H23N
Molecular Weight: 313.87
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 242


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 9-(gamma-Methylaminopropyl)-9,10-dihydro-9,10-ethanoanthracene Hydrochloride

Formula: C₂₀H₂₃N
MW: 313.87 g/mol
Melting Pt: 242 °C
MDL Number: MFCD00079464
CAS Number: 10347-81-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ccb56b84a956fd43c26e4d3ec02fa10e4840fb63&vid=ccb56b84a956fd43c26e4d3ec02fa10e4840fb63&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR