Monomethyl-trans-1,4-cyclohexanedicarboxylate ≥97.0% (by GC, titration analysis)
Supplier: TCI America
M2526-25G
M2526-5G
TCM2526-25GEA
278.08
CAD
TCM2526-25G
TCM2526-5G
Monomethyl-trans-1,4-cyclohexanedicarboxylate ≥97.0% (by GC, titration analysis)
Monomethyl-trans-1,4-cyclohexanedicarboxylate
CAS Number: 15177-67-0
MDL Number: MFCD01311247
Molecular Formula: C9H14O4
Molecular Weight: 186.21
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 128
Other Product Names / Synonyms: trans-1,4-Cyclohexanedicarboxylic Acid Monomethyl Ester, Methyl Hydrogen trans-1,4-Cyclohexanedicarboxylate
Formula:
C₉H₁₄O₄ Melting Pt: 128 °C |
MDL Number:
MFCD01311247 CAS Number: 15177-67-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...