Methyl azetidine-3-carboxylate hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Azetidine-3-carboxylic acid methyll ester hydrochloride, Methyl Azetidine-3-carboxylate Hydrochloride, Azetidine-3-carboxylic methyl ester hydrochloride

M2521-1G M2521-5G
TCM2521-1GEA 50.56 CAD
TCM2521-1G TCM2521-5G
Methyl azetidine-3-carboxylate hydrochloride ≥98.0% (by titrimetric analysis)
Methyl azetidine-3-carboxylate hydrochloride

CAS Number: 100202-39-9
MDL Number: MFCD01861758
Molecular Formula: C5H9NO2
Molecular Weight: 151.59
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 192
Melting point (°C): 95
Flash Point (°C): 69


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: Azetidine-3-carboxylic Acid Methyl Ester Hydrochloride

Formula: C₅H₁₀ClNO₂
MW: 151.59 g/mol
Boiling Pt: 192 °C
Melting Pt: 95 °C
Flash Pt: 69 °C
MDL Number: MFCD01861758
CAS Number: 100202-39-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0dbff142e68a5ee2e0fd180087eafe802311f6ca&vid=0dbff142e68a5ee2e0fd180087eafe802311f6ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR