Methyl caffeate ≥98.0%

Supplier: TCI America

Synonyms: 3,4-Dihydroxycinnamic acid methyl ester, Methyl 3,4-dihydroxycinnamate, Caffeic acid methyl ester, Methyl 3-(3,4-dihydroxyphenyl)acrylate

M2519-1G M2519-200MG
TCM2519-1GEA 225.94 CAD
TCM2519-1G TCM2519-200MG
Methyl caffeate ≥98.0%
Methyl caffeate

CAS Number: 3843-74-1
MDL Number: MFCD00210468
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 160
Lambda max.: 328 nm (MeOH)


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Antioxidants, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: Caffeic Acid Methyl Ester, Methyl 3,4-Dihydroxycinnamate, 3,4-Dihydroxycinnamic Acid Methyl Ester

Formula: C₁₀H₁₀O₄
Melting Pt: 160 °C
MDL Number: MFCD00210468
CAS Number: 3843-74-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdf80865fe00fe0ee54f74e38a2dec6ff28669d9&vid=bdf80865fe00fe0ee54f74e38a2dec6ff28669d9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR