6'-Methoxy-2,3'-bipyridine ≥97.0% (by GC)

Supplier: TCI America

M2509-1G M2509-200MG
TCM2509-1GEA 778.47 CAD
TCM2509-1G TCM2509-200MG
6'-Methoxy-2,3'-bipyridine ≥97.0% (by GC)
2-Methoxy-5-(pyridin-2-yl)pyridine

CAS Number: 381725-49-1
MDL Number: MFCD11707142
Molecular Formula: C11H10N2O
Molecular Weight: 186.21
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 104


Other Product Names / Synonyms: 6'-Methoxy-2,3'-bipyridine

Formula: C₁₁H₁₀N₂O
Boiling Pt: 104 °C
MDL Number: MFCD11707142
CAS Number: 381725-49-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36a982a27f13b1e624dde390457362079861ca29&vid=36a982a27f13b1e624dde390457362079861ca29&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR