Methyl-4-methoxypicolinate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M2508-1G
M2508-5G
TCM2508-1GEA
126.4
CAD
TCM2508-1G
TCM2508-5G
Methyl-4-methoxypicolinate ≥98.0% (by GC, titration analysis)
Methyl 4-methoxypicolinate
CAS Number: 29681-43-4
MDL Number: MFCD00956223
Molecular Formula: C8H9NO3
Molecular Weight: 167.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 52
Other Product Names / Synonyms: 4-Methoxypyridine-2-carboxylic Acid Methyl Ester, Methyl 4-Methoxypicolinate, 4-Methoxypicolinic Acid Methyl Ester
Formula:
C₈H₉NO₃ Melting Pt: 52 °C |
MDL Number:
MFCD00956223 CAS Number: 29681-43-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...