4-Methylpiperazine-1-ethanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M2499-25G M2499-5G
TCM2499-25GEA 186.44 CAD
TCM2499-25G TCM2499-5G
4-Methylpiperazine-1-ethanol ≥98.0% (by GC, titration analysis)
4-Methylpiperazine-1-ethanol

CAS Number: 5464-12-0
MDL Number: MFCD03840691
Molecular Formula: C7H16N2O
Molecular Weight: 144.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 55
Melting point (°C): 41


Other Product Names / Synonyms: 1-(2-Hydroxyethyl)-4-methylpiperazine

Formula: C₇H₁₆N₂O
Boiling Pt: 55 °C
Melting Pt: 41 °C
MDL Number: MFCD03840691
CAS Number: 5464-12-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d4b20db0b3b52fc73115d8c35f8f0830d14e59d&vid=7d4b20db0b3b52fc73115d8c35f8f0830d14e59d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR