1-Methyl-1H-pyrazole-5-carboxylic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

M2494-1G M2494-5G
TCM2494-1GEA 50.56 CAD
TCM2494-1G TCM2494-5G
1-Methyl-1H-pyrazole-5-carboxylic acid ≥97.0% (by GC, titration analysis)
1-Methyl-1H-pyrazole-5-carboxylic acid

CAS Number: 16034-46-1
MDL Number: MFCD00464253
Molecular Formula: C5H6N2O2
Molecular Weight: 126.12
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 225
Lambda max.: 259 nm (MeOH)

Formula: C₅H₆N₂O₂
Melting Pt: 225 °C
MDL Number: MFCD00464253
CAS Number: 16034-46-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c34c88a7e379407a44b05951bfcabf12b5d54eeb&vid=c34c88a7e379407a44b05951bfcabf12b5d54eeb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR