4-Methyl-3-cyclohexene-1-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Methylcyclohex-3-enecarboxylic acid

M2492-1G M2492-5G
TCM2492-1GEA 146.94 CAD
TCM2492-1G TCM2492-5G
4-Methyl-3-cyclohexene-1-carboxylic acid ≥98.0% (by GC, titration analysis)
4-Methyl-3-cyclohexene-1-carboxylic acid

CAS Number: 4342-60-3
Molecular Formula: C8H12O2
Molecular Weight: 140.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 100

Formula: C₈H₁₂O₂
MW: 140.18 g/mol
Melting Pt: 100 °C
MDL Number: MFCD02238356
CAS Number: 4342-60-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2bab371f159a8f3c91f0d87fdb623f11360423fb&vid=2bab371f159a8f3c91f0d87fdb623f11360423fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR