2-Methyl-5-oxo-1-cyclopentenyl butyrate ≥98.0%

Supplier: TCI America

Synonyms: 2-Methyl-5-oxocyclopent-1-en-1-yl butyrate, Butyric Acid 2-Methyl-5-oxo-1-cyclopentenyl Ester

M2486-25G M2486-5G
TCM2486-25GEA 781.78 CAD
TCM2486-25G TCM2486-5G
2-Methyl-5-oxo-1-cyclopentenyl butyrate ≥98.0%
2-Methyl-5-oxo-1-cyclopentenyl butyrate

CAS Number: 68227-51-0
MDL Number: MFCD00798052
Molecular Formula: C10H14O3
Molecular Weight: 182.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: Butyric Acid 2-Methyl-5-oxo-1-cyclopentenyl Ester

Formula: C₁₀H₁₄O₃
MW: 182.22 g/mol
Boiling Pt: 85 °C
Density: 1.06 (20 °C)
MDL Number: MFCD00798052
CAS Number: 68227-51-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=728216a8d069c7165711b2ab33d9a2a4c5b2f4f0&vid=728216a8d069c7165711b2ab33d9a2a4c5b2f4f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR