2-(4-Methyl-2-thioxo-2,3-dihydrothiazol-5-yl)acetic acid ≥98.0%

Supplier: TCI America

Synonyms: 5-(Carboxymethyl)-2-mercapto-4-methylthiazole, (2-Mercapto-4-methyl-5-thiazolyl)acetic acid

M2481-25G M2481-5G
TCM2481-25GEA 79 CAD
TCM2481-25G TCM2481-5G
2-(4-Methyl-2-thioxo-2,3-dihydrothiazol-5-yl)acetic acid ≥98.0%
2-(4-Methyl-2-thioxo-2,3-dihydrothiazol-5-yl)acetic acid

CAS Number: 34272-64-5
MDL Number: MFCD01750430
Molecular Formula: C6H7NO2S2
Molecular Weight: 189.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 211
Lambda max.: 320 nm (H2O)


Other Product Names / Synonyms: 5-(Carboxymethyl)-2-mercapto-4-methylthiazole

Formula: C₆H₇NO₂S₂
Melting Pt: 211 °C
MDL Number: MFCD01750430
CAS Number: 34272-64-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ca8fb962916e96d6f9d0c06f0a6ab986fda2d56&vid=4ca8fb962916e96d6f9d0c06f0a6ab986fda2d56&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR