6-Methylnicotinamide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

M2477-25G M2477-5G
TCM2477-25GEA 783.52 CAD
TCM2477-25G TCM2477-5G
6-Methylnicotinamide ≥98.0% (by HPLC, titration analysis)
6-Methylnicotinamide

CAS Number: 6960-22-1
MDL Number: MFCD00006342
Molecular Formula: C7H8N2O
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 199


Other Product Names / Synonyms: 2-Methylpyridine-5-carboxamide, 6-Methylpyridine-3-carboxamide

Formula: C₇H₈N₂O
Melting Pt: 199 °C
MDL Number: MFCD00006342
CAS Number: 6960-22-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95fd10d504cc155aa85acf65de3c39627989c812&vid=95fd10d504cc155aa85acf65de3c39627989c812&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR