Methyl-2-(trifluoromethyl)benzoate ≥98.0% (by GC)

Supplier: TCI America

M2475-25G M2475-5G
TCM2475-25GEA 146.94 CAD
TCM2475-25G TCM2475-5G
Methyl-2-(trifluoromethyl)benzoate ≥98.0% (by GC)
Methyl 2-(trifluoromethyl)benzoate

CAS Number: 344-96-7
MDL Number: MFCD00236050
Molecular Formula: C9H7F3O2
Molecular Weight: 204.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Specific Gravity (20/20): 1.31


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-(Trifluoromethyl)benzoic Acid Methyl Ester, Methyl alpha,alpha,alpha-Trifluoro-o-toluate

Formula: C₉H₇F₃O₂
Boiling Pt: 105 °C
Density: 1.31 (20 °C)
MDL Number: MFCD00236050
CAS Number: 344-96-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e80eed19e42e5ff3aeb9fbb022b68b6fc73282a&vid=5e80eed19e42e5ff3aeb9fbb022b68b6fc73282a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR