2-Methylene-1,3-propanediol ≥96.0% (by GC)

Supplier: TCI America

M2473-1G M2473-5G
TCM2473-1GEA 57.83 CAD
TCM2473-1G TCM2473-5G
2-Methylene-1,3-propanediol ≥96.0% (by GC)
2-Methylene-1,3-propanediol

CAS Number: 3513-81-3
MDL Number: MFCD00075162
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Flash Point (°C): 110
Specific Gravity (20/20): 1.07

Formula: C₄H₈O₂
Boiling Pt: 95 °C
Density: 1.07 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00075162
CAS Number: 3513-81-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06ac1c28fd4c8621c43b9e4b0e870249675d342e&vid=06ac1c28fd4c8621c43b9e4b0e870249675d342e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR