2-(3-Methoxyphenethyl)phenol ≥98.0%
Supplier: TCI America
Synonyms:
2-[2-(3-Methoxyphenyl)ethyl]phenol
M2472-1G
M2472-5G
TCM2472-1GEA
117.39
CAD
TCM2472-1G
TCM2472-5G
2-(3-Methoxyphenethyl)phenol ≥98.0%
2-(3-Methoxyphenethyl)phenol
CAS Number: 167145-13-3
MDL Number: MFCD09800498
Molecular Formula: C15H16O2
Molecular Weight: 228.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 47
Other Product Names / Synonyms: 2-[2-(3-Methoxyphenyl)ethyl]phenol
Formula:
C₁₅H₁₆O₂ MW: 228.29 g/mol Melting Pt: 47 °C |
MDL Number:
MFCD09800498 CAS Number: 167145-13-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...