Methyl-5-methylpyrazole-3-carboxylate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 5-methyl-1H-pyrazole-3-carboxylate, Methyl 5-methyl[1H]pyrazole-3-carboxylate, 5-Methylpyrazole-3-carboxylic Acid Methyl Ester

M2465-1G M2465-5G
TCM2465-1GEA 145.52 CAD
TCM2465-1G TCM2465-5G
Methyl-5-methylpyrazole-3-carboxylate ≥98.0%
Methyl-5-methylpyrazole-3-carboxylate

CAS Number: 25016-17-5
MDL Number: MFCD03778987
Molecular Formula: C6H8N2O2
Molecular Weight: 140.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 83


Other Product Names / Synonyms: 5-Methylpyrazole-3-carboxylic Acid Methyl Ester

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Melting Pt: 83 °C
MDL Number: MFCD03778987
CAS Number: 25016-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f92d75d5565d6bd5ba046853d2f26e406b274b9&vid=9f92d75d5565d6bd5ba046853d2f26e406b274b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR