(3-Methoxybenzyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
M2463-1G
M2463-5G
TCM2463-1GEA
82.16
CAD
TCM2463-1G
TCM2463-5G
(3-Methoxybenzyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
(3-Methoxybenzyl)triphenylphosphonium chloride
CAS Number: 18880-05-2
MDL Number: MFCD09836189
Molecular Formula: C26H24ClOP
Molecular Weight: 418.90
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 268
Formula:
C₂₆H₂₄ClOP Melting Pt: 268 °C |
MDL Number:
MFCD09836189 CAS Number: 18880-05-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...