Methyl-2-bromoisonicotinate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 2-bromoisonicotinate

M2461-1G M2461-5G
TCM2461-1GEA 180.12 CAD
TCM2461-1G TCM2461-5G
Methyl-2-bromoisonicotinate ≥98.0%
Methyl 2-bromoisonicotinate

CAS Number: 26156-48-9
MDL Number: MFCD03791265
Molecular Formula: C7H6BrNO2
Molecular Weight: 216.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 37


Other Product Names / Synonyms: 2-Bromoisonicotinic Acid Methyl Ester, Methyl 2-Bromopyridine-4-carboxylate, 2-Bromopyridine-4-carboxylic Acid Methyl Ester

Formula: C₇H₆BrNO₂
Melting Pt: 37 °C
MDL Number: MFCD03791265
CAS Number: 26156-48-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bc45f3cb7e2d03482f486f93df43b5e9a57c502&vid=9bc45f3cb7e2d03482f486f93df43b5e9a57c502&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR