4-Methyl-1-naphthaleneboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
M2457-1G
M2457-5G
TCM2457-1GEA
66.36
CAD
TCM2457-1G
TCM2457-5G
4-Methyl-1-naphthaleneboronic acid (contains varying amounts of Anhydride)
4-Methyl-1-naphthaleneboronic acid
CAS Number: 103986-53-4
MDL Number: MFCD01632204
Molecular Formula: C11H11BO2
Molecular Weight: 186.02
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 207
Other Product Names / Synonyms: 4-Methyl-1-naphthylboronic Acid
Formula:
C₁₁H₁₁BO₂ Melting Pt: 207 °C |
MDL Number:
MFCD01632204 CAS Number: 103986-53-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...