2-(Methylthio)benzo[d]oxazole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-(Methylthio)benzoxazole

M2454-5G M2454-1G
TCM2454-5GEA 339.7 CAD
TCM2454-5G 76220-236
2-(Methylthio)benzo[d]oxazole ≥98.0% (by GC)
2-(Methylthio)benzo[d]oxazole

CAS Number: 13673-62-6
MDL Number: MFCD00160027
Molecular Formula: C8H7NOS
Molecular Weight: 165.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Freezing point (°C): 9
Specific Gravity (20/20): 1.24
Lambda max.: 278 nm (MeOH)

Formula: C₈H₇NOS
MW: 165.2100 g/mol
Boiling Pt: 122 °C
Density: 1.24 (20 °C)
MDL Number: MFCD00160027
CAS Number: 13673-62-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=317b7866d9326b1b27c96654999f6937fcf51a2d&vid=317b7866d9326b1b27c96654999f6937fcf51a2d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR