Methyl-2-bromonicotinate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 2-bromonicotinate, Methyl 2-bromopyridine-3-carboxylate

M2449-1G M2449-5G
TCM2449-1GEA 139.04 CAD
TCM2449-1G TCM2449-5G
Methyl-2-bromonicotinate ≥98.0%
Methyl 2-bromonicotinate

CAS Number: 52718-95-3
MDL Number: MFCD07367915
Molecular Formula: C7H6BrNO2
Molecular Weight: 216.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Freezing point (°C): 31


Other Product Names / Synonyms: 2-Bromonicotinic Acid Methyl Ester, Methyl 2-Bromopyridine-3-carboxylate, 2-Bromopyridine-3-carboxylic Acid Methyl Ester

Formula: C₇H₆BrNO₂
MDL Number: MFCD07367915
CAS Number: 52718-95-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c729eb6fe0e7d3452f43e8979f68ef08c6b17d7f&vid=c729eb6fe0e7d3452f43e8979f68ef08c6b17d7f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR