Methyl-3-hydroxyisoxazole-5-carboxylate ≥98.0% (by GC)

Supplier: TCI America

M2441-1G M2441-5G
TCM2441-1GEA 74.42 CAD
TCM2441-1G TCM2441-5G
Methyl-3-hydroxyisoxazole-5-carboxylate ≥98.0% (by GC)
Methyl-3-hydroxyisoxazole-5-carboxylate

CAS Number: 10068-07-2
MDL Number: MFCD00062870
Molecular Formula: C5H5NO4
Molecular Weight: 143.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 164


Other Product Names / Synonyms: 3-Hydroxyisoxazole-5-carboxylic Acid Methyl Ester

Formula: C₅H₅NO₄
Melting Pt: 164 °C
MDL Number: MFCD00062870
CAS Number: 10068-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=801de258b4cbc9c8519e102481ff4eaed4d5a33a&vid=801de258b4cbc9c8519e102481ff4eaed4d5a33a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR