5-Methoxy-1H-indole-3-carboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: 5-Methoxyindole-3-carboxylic acid

M2438-1G M2438-5G
TCM2438-1GEA 115.66 CAD
TCM2438-1G TCM2438-5G
5-Methoxy-1H-indole-3-carboxylic acid ≥98.0%
5-Methoxy-1H-indole-3-carboxylic acid

CAS Number: 10242-01-0
MDL Number: MFCD03265451
Molecular Formula: C10H9NO3
Molecular Weight: 191.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 173

Formula: C10H9NO3
Melting Pt: 173 °C
MDL Number: MFCD03265451
CAS Number: 10242-01-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e11f5cc7c84535ff2ed55a855c6ef2ef3864eb5&vid=1e11f5cc7c84535ff2ed55a855c6ef2ef3864eb5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR