N-Methyl-L-alanine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: H-N-Me-Ala-OH, N-Me-Ala-OH

M2436-1G M2436-5G
TCM2436-1GEA 75.84 CAD
TCM2436-1G TCM2436-5G
N-Methyl-L-alanine ≥98.0% (by titrimetric analysis)
N-Methyl-L-alanine

CAS Number: 3913-67-5
MDL Number: MFCD00037241
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 300
Specific rotation [a]20/D: 11 deg (C=10, 6mol/L HCl)

Formula: C₄H₉NO₂
MW: 103.12 g/mol
Melting Pt: 300 °C
MDL Number: MFCD00037241
CAS Number: 3913-67-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=856453810e4b1efff3400c4244eb5e5cc8ebaa0d&vid=856453810e4b1efff3400c4244eb5e5cc8ebaa0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR