1-Methylxanthine ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3,7-Dihydro-1-methyl-1H-purine-2,6-dione, 2,6-Dihydroxy-1-methylpurine, 1-Methyl-1H-purine-2,6(3H,7H)-dione

M2432-200MG M2432-50MG
TCM2432-200MGEA 372.88 CAD
TCM2432-200MG TCM2432-50MG
1-Methylxanthine ≥97.0% (by HPLC, titration analysis)
1-Methylxanthine

CAS Number: 6136-37-4
MDL Number: MFCD00005561
Molecular Formula: C6H6N4O2
Molecular Weight: 166.14
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Lambda max.: 270 nm (H2O)

Formula: C₆H₆N₄O₂
MW: 166.14
MDL Number: MFCD00005561
CAS Number: 6136-37-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c165abbe112a2923daaa13cbd5239c847c55b479&vid=c165abbe112a2923daaa13cbd5239c847c55b479&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR