Methyl-5-methylnicotinate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M2428-1G
M2428-5G
TCM2428-1GEA
45.82
CAD
TCM2428-1G
TCM2428-5G
Methyl-5-methylnicotinate ≥98.0% (by GC, titration analysis)
Methyl 5-methylnicotinate
CAS Number: 29681-45-6
MDL Number: MFCD01075739
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 90
Melting point (°C): 48
Other Product Names / Synonyms: 5-Methylnicotinic Acid Methyl Ester, Methyl 5-Methylpyridine-3-carboxylate, 5-Methylpyridine-3-carboxylic Acid Methyl Ester
Formula:
C₈H₉NO₂ Boiling Pt: 90 °C Melting Pt: 48 °C |
MDL Number:
MFCD01075739 CAS Number: 29681-45-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...