4-Mercaptophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
M2418-1G
M2418-5G
TCM2418-1GEA
284.57
CAD
TCM2418-1G
TCM2418-5G
4-Mercaptophenylboronic acid (contains varying amounts of Anhydride)
4-Mercaptophenylboronic acid
CAS Number: 237429-33-3
MDL Number: MFCD03093887
Molecular Formula: C6H7BO2S
Molecular Weight: 153.99
Form: Crystal
Color: White
Melting point (°C): 220
Other Product Names / Synonyms: 4-Mercaptobenzeneboronic Acid
Formula:
C₆H₇BO₂S Melting Pt: 220 °C |
MDL Number:
MFCD03093887 CAS Number: 237429-33-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...