erythro-3-Methoxy-2-methyl-4-phenylbutyric acid ≥98.0%

Supplier: TCI America

M2415-100MG M2415-500MG
TCM2415-100MGEA 341.88 CAD
TCM2415-100MG TCM2415-500MG
erythro-3-Methoxy-2-methyl-4-phenylbutyric acid ≥98.0%
erythro-3-Methoxy-2-methyl-4-phenylbutyric acid

Molecular Weight: 208.26
Purity/Analysis Method: >98.0% (GC)

Storage Temperature: <0°C
Molecular Formula: C12H16O3

Formula: C₁₂H₁₆O₃
MW: 208.26 g/mol
CAS Number: 833483-97-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb1c8ea35376e5b68e5d857e0c392b90d5d59d32&vid=fb1c8ea35376e5b68e5d857e0c392b90d5d59d32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR