Fumaric acid monomethyl ester ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (E)-4-methoxy-4-oxobut-2-enoic acid, Monomethyl Fumarate, Fumaric acid monomethyl ester, Methyl Hydrogen Fumarate

M2413-5G M2413-25G
TCM2413-5GEA 67.78 CAD
TCM2413-5G TCM2413-25G
Fumaric acid monomethyl ester ≥98.0% (by GC)
Fumaric acid monomethyl ester

CAS Number: 2756-87-8
MDL Number: MFCD00063174
Molecular Formula: C5H6O4
Molecular Weight: 130.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 147
Lambda max.: 209 nm (MeOH)


Other Product Names / Synonyms: Methyl Hydrogen Fumarate, Fumaric Acid Monomethyl Ester

Formula: C₅H₆O₄
MW: 130.1 g/mol
Melting Pt: 147 °C
MDL Number: MFCD00063174
CAS Number: 2756-87-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78266193d5331d99656099d7d3cab60dc263e10e&vid=78266193d5331d99656099d7d3cab60dc263e10e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR