4-Methyl-2-hexylamine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M2409-5G M2409-1G
TCM2409-5GEA 133.83 CAD
TCM2409-5G TCM2409-1G
4-Methyl-2-hexylamine hydrochloride ≥98.0% (by titrimetric analysis)
4-Methyl-2-hexylamine hydrochloride

CAS Number: 13803-74-2
MDL Number: MFCD09038354
Molecular Formula: C7H17N
Molecular Weight: 151.68
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 130


Other Product Names / Synonyms: Methylhexanamine Hydrochloride, 1,3-Dimethylpentylamine Hydrochloride, 1,3-Dimethylamylamine Hydrochloride, 2-Amino-4-methylhexane Hydrochloride

Formula: C₇H₁₇N
Melting Pt: 130 °C
MDL Number: MFCD09038354
CAS Number: 13803-74-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e75852d57b3a94841a88ebd36fb8f1c43ed9fb1&vid=9e75852d57b3a94841a88ebd36fb8f1c43ed9fb1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR