Mecarbinate ≥98.0% (by GC)

Supplier: TCI America

M2407-1G M2407-5G
TCM2407-1GEA 45.82 CAD
TCM2407-1G TCM2407-5G
Mecarbinate ≥98.0% (by GC)
Mecarbinate

CAS Number: 15574-49-9
MDL Number: MFCD00451373
Molecular Formula: C13H15NO3
Molecular Weight: 233.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 212


Other Product Names / Synonyms: Ethyl 5-Hydroxy-1,2-dimethylindole-3-carboxylate, 5-Hydroxy-1,2-dimethylindole-3-carboxylic Acid Ethyl Ester

Formula: C₁₃H₁₅NO₃
Melting Pt: 212 °C
MDL Number: MFCD00451373
CAS Number: 15574-49-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd3494c81267a9f2e40a7eca20c773af2bc2df95&vid=fd3494c81267a9f2e40a7eca20c773af2bc2df95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR