Methyl-3-oxo-4-phenylbutyrate (mixture of isomers) ≥96.0%

Supplier: TCI America

Synonyms: Methyl 3-oxo-4-phenylbutanoate, Methyl 4-Phenylacetoacetate, 3-Oxo-4-phenylbutyric Acid Methyl Ester, 4-Phenylacetoacetic Acid Methyl Ester

M2396-1G M2396-5G
TCM2396-1GEA 225.94 CAD
TCM2396-1G TCM2396-5G
Methyl-3-oxo-4-phenylbutyrate (mixture of isomers) ≥96.0%
Methyl-3-oxo-4-phenylbutyrate (mixture of isomers)

CAS Number: 37779-49-0
MDL Number: MFCD03424746
Molecular Formula: C11H12O3
Molecular Weight: 192.21
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 136
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: 3-Oxo-4-phenylbutyric Acid Methyl Ester, Methyl 4-Phenylacetoacetate, 4-Phenylacetoacetic Acid Methyl Ester

Formula: C₁₁H₁₂O₃
Boiling Pt: 136 °C
Density: 1.13 (20 °C)
MDL Number: MFCD03424746
CAS Number: 37779-49-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=649399436d1edaa3257049d5e058fed7158af4d5&vid=649399436d1edaa3257049d5e058fed7158af4d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR