Methyl-2-chloro-L-mandelate ≥98.0% (by GC)

Supplier: TCI America

M2383-1G M2383-5G
TCM2383-1GEA 223.1 CAD
TCM2383-1G TCM2383-5G
Methyl-2-chloro-L-mandelate ≥98.0% (by GC)
Methyl-2-chloro-L-mandelate

CAS Number: 32345-60-1
Molecular Formula: C9H9ClO3
Molecular Weight: 200.62
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.31
Specific rotation [a]20/D: 180 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: 2-Chloro-L-mandelic Acid Methyl Ester, Methyl (S)-2'-Chloro-alpha-hydroxyphenylacetate, (S)-2'-Chloro-alpha-hydroxyphenylacetic Acid Methyl Ester

Formula: C₉H₉ClO₃
Density: 1.31 (20 °C)
MDL Number: MFCD02259731
CAS Number: 32345-60-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b93414d12a352180908a1ca877bc0526128fef65&vid=b93414d12a352180908a1ca877bc0526128fef65&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR