Methyl-2-chloro-D-mandelate ≥98.0%

Supplier: TCI America

Synonyms: Methyl (R)-2'-Chloro-alpha-hydroxyphenylacetate, (R)-Methyl 2-(2-chlorophenyl)-2-hydroxyacetate, 2-Chloro-D-mandelic Acid Methyl Ester, (R)-2'-Chloro-alpha-hydroxyphenylacetic Acid Methyl Ester

M2382-25G M2382-5G
TCM2382-25GEA 580.18 CAD
TCM2382-25G TCM2382-5G
Methyl-2-chloro-D-mandelate ≥98.0%
Methyl-2-chloro-D-mandelate

CAS Number: 32345-59-8
MDL Number: MFCD02259751
Molecular Formula: C9H9ClO3
Molecular Weight: 200.62
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 34
Specific rotation [a]20/D: -180 deg (C=0.4, CHCl3)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: 2-Chloro-D-mandelic Acid Methyl Ester, Methyl (R)-2'-Chloro-alpha-hydroxyphenylacetate, (R)-2'-Chloro-alpha-hydroxyphenylacetic Acid Methyl Ester

Formula: C₉H₉ClO₃
MW: 200.62 g/mol
Melting Pt: 34 °C
MDL Number: MFCD02259751
CAS Number: 32345-59-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c723970d612026e2724523b59e6b7f8b1eeef91d&vid=c723970d612026e2724523b59e6b7f8b1eeef91d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR