3-Methyl-2-furoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M2376-1G M2376-5G
TCM2376-1GEA 83.74 CAD
TCM2376-1G TCM2376-5G
3-Methyl-2-furoic acid ≥98.0% (by GC, titration analysis)
3-Methyl-2-furoic acid

CAS Number: 4412-96-8
MDL Number: MFCD00014111
Molecular Formula: C6H6O3
Molecular Weight: 126.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 136


Other Product Names / Synonyms: 3-Methyl-2-furoic Acid, 3-Methylpyromucic Acid

Formula: C₆H₆O₃
Melting Pt: 136 °C
MDL Number: MFCD00014111
CAS Number: 4412-96-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e9eecbd9a9f46ed2464cd7ace10e9e48744ec7e&vid=7e9eecbd9a9f46ed2464cd7ace10e9e48744ec7e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR