1-Methyl-1H-pyrazol-4-carbaldehyde ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Methyl-1H-pyrazole-4-carbaldehyde, 1-Methyl-1H-pyrazole-4-carboxaldehyde

M2372-1G M2372-5G
TCM2372-1GEA 42.66 CAD
TCM2372-1G TCM2372-5G
1-Methyl-1H-pyrazol-4-carbaldehyde ≥98.0% (by GC)
1-Methyl-1H-pyrazol-4-carbaldehyde

CAS Number: 25016-11-9
MDL Number: MFCD00460465
Molecular Formula: C5H6N2O
Molecular Weight: 110.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 81
Melting point (°C): 33
Flash Point (°C): 110


Other Product Names / Synonyms: 4-Formyl-1-methylpyrazole

Formula: C₅H₆N₂O
MW: 110.12 g/mol
Boiling Pt: 81 °C
Melting Pt: 33 °C
Flash Pt: 110 °C
MDL Number: MFCD00460465
CAS Number: 25016-11-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7e379a5532c925e64a65c6e5345a5a018f28fa9&vid=c7e379a5532c925e64a65c6e5345a5a018f28fa9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR