3-Methylflavone-8-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

M2371-25G M2371-5G
TCM2371-25GEA 175.38 CAD
TCM2371-25G TCM2371-5G
3-Methylflavone-8-carboxylic acid ≥98.0% (by HPLC, titration analysis)
3-Methylflavone-8-carboxylic acid

CAS Number: 3468-01-7
MDL Number: MFCD00040998
Molecular Formula: C17H12O4
Molecular Weight: 280.28
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 236


Product Categories: Bioscience, Aromatic Polyketides
Other Product Names / Synonyms: 3-Methyl-2-phenylchromone-8-carboxylic Acid

Formula: C₁₇H₁₂O₄
Melting Pt: 236 °C
MDL Number: MFCD00040998
CAS Number: 3468-01-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da4448a08871b6e5fca5f9198763f2137488ab8b&vid=da4448a08871b6e5fca5f9198763f2137488ab8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR