Methyl-1,2,4-triazole-3-carboxylate ≥98.0%
Supplier: TCI America
Synonyms:
Methyl 1H-1,2,4-triazole-3-carboxylate, Methyl 1H-1,2,4-triazole-5-carboxylate
M2368-25G
M2368-5G
TCM2368-25GEA
140.62
CAD
TCM2368-25G
TCM2368-5G
Methyl-1,2,4-triazole-3-carboxylate ≥98.0%
Methyl 1,2,4-triazole-3-carboxylate
CAS Number: 4928-88-5
MDL Number: MFCD00135989
Molecular Formula: C4H5N3O2
Molecular Weight: 127.10
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 189
Other Product Names / Synonyms: 1,2,4-Triazole-3-carboxylic Acid Methyl Ester
Formula:
C₄H₅N₃O₂ Melting Pt: 189 °C |
MDL Number:
MFCD00135989 CAS Number: 4928-88-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...