5-Methoxyflavone ≥98.0% (by GC)

Supplier: TCI America

M2365-1G M2365-5G
TCM2365-1GEA 145.36 CAD
TCM2365-1G TCM2365-5G
5-Methoxyflavone ≥98.0% (by GC)
5-Methoxyflavone

CAS Number: 42079-78-7
MDL Number: MFCD00016942
Molecular Formula: C16H12O3
Molecular Weight: 252.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 132

Formula: C₁₆H₁₂O₃
Melting Pt: 132 °C
MDL Number: MFCD00016942
CAS Number: 42079-78-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46425a95f2af0c296b5f5361d6dae2c41fc0ba18&vid=46425a95f2af0c296b5f5361d6dae2c41fc0ba18&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR