3-Methoxyflavone ≥98.0% (by GC)

Supplier: TCI America

M2364-1G M2364-5G
TCM2364-1GEA 159.58 CAD
TCM2364-1G TCM2364-5G
3-Methoxyflavone ≥98.0% (by GC)
3-Methoxyflavone

CAS Number: 7245-02-5
MDL Number: MFCD00017612
Molecular Formula: C16H12O3
Molecular Weight: 252.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 114
Lambda max.: 299 nm (EtOH)

Formula: C₁₆H₁₂O₃
Melting Pt: 114 °C
MDL Number: MFCD00017612
CAS Number: 7245-02-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f804413a51d0a00e2fdb7efaae1b6772e0acb9c&vid=0f804413a51d0a00e2fdb7efaae1b6772e0acb9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR