3-Methyl-6-nitro-1H-indazole ≥98.0% (by GC)

Supplier: TCI America

M2358-25G M2358-5G
TCM2358-25GEA 137.46 CAD
TCM2358-25G TCM2358-5G
3-Methyl-6-nitro-1H-indazole ≥98.0% (by GC)
3-Methyl-6-nitro-1H-indazole

CAS Number: 6494-19-5
MDL Number: MFCD07082704
Molecular Formula: C8H7N3O2
Molecular Weight: 177.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 186

Formula: C₈H₇N₃O₂
Melting Pt: 186 °C
MDL Number: MFCD07082704
CAS Number: 6494-19-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73eb4a65c8c98efcab6a2d051d0f8445f0307bbd&vid=73eb4a65c8c98efcab6a2d051d0f8445f0307bbd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR