2-Formyl-6-methoxypyridine ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 6-Methoxypicolinaldehyde, 6-Methoxypyridine-2-carboxaldehyde, 6-Methoxypyridine-2-carbaldehyde

M2357-1G M2357-5G
TCM2357-1GEA 88.48 CAD
TCM2357-1G TCM2357-5G
2-Formyl-6-methoxypyridine ≥97.0% (by GC)
2-Formyl-6-methoxypyridine

CAS Number: 54221-96-4
MDL Number: MFCD05664030
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): 81
Specific Gravity (20/20): 1.16


Other Product Names / Synonyms: 6-Methoxypicolinaldehyde, 2-Formyl-6-methoxypyridine

Formula: C₇H₇NO₂
MW: 137.14
Boiling Pt: 104 °C
Density: 1.16 (20 °C)
Flash Pt: 81 °C
MDL Number: MFCD05664030
CAS Number: 54221-96-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6257f499430e83e8a605d9ed66c9065a752a6547&vid=6257f499430e83e8a605d9ed66c9065a752a6547&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR