2-Methylthio-4-pyrimidinol ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-(methylsulfanyl)-3,4-dihydropyrimidin-4-one, 2-(Methylthio)pyrimidin-4(3H)-one, 2-(methylsulphanyl)-3,4-dihydropyrimidin-4-one, 2-Methylthio-4-pyrimidone, 4-Hydroxy-2-methylthiopyrimidine, S-Methyl-2-thiouracil

M2355-1G M2355-5G
TCM2355-1GEA 30.02 CAD
TCM2355-1G TCM2355-5G
2-Methylthio-4-pyrimidinol ≥98.0% (by HPLC, titration analysis)
2-Methylthio-4-pyrimidinol

CAS Number: 5751-20-2
MDL Number: MFCD00047373
Molecular Formula: C5H6N2OS
Molecular Weight: 142.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 202


Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: S-Methyl-2-thiouracil, 4-Hydroxy-2-methylthiopyrimidine

Formula: C₅H₆N₂OS
MW: 142.18 g/mol
Melting Pt: 202 °C
MDL Number: MFCD00047373
CAS Number: 5751-20-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42e43b51316facc22e49af41c0d55678bb5bab8f&vid=42e43b51316facc22e49af41c0d55678bb5bab8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR