Methyl-1-aminocyclopentanecarboxylate hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 1-Aminocyclopentanecarboxylic Acid Methyl Ester Hydrochloride, Methyl 1-aminocyclopentanecarboxylate hydrochloride, Methyl 1-Aminocyclopentanecarboxylate HCl, Cycloleucine Methyl Ester Hydrochloride

M2351-1G M2351-5G
TCM2351-1GEA 63.2 CAD
TCM2351-1G TCM2351-5G
Methyl-1-aminocyclopentanecarboxylate hydrochloride ≥98.0%
Methyl-1-aminocyclopentanecarboxylate hydrochloride

CAS Number: 60421-23-0
MDL Number: MFCD03840364
Molecular Formula: C7H13NO2
Molecular Weight: 179.64
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 203


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: 1-Aminocyclopentanecarboxylic Acid Methyl Ester Hydrochloride, Cycloleucine Methyl Ester Hydrochloride

Formula: C₇H₁₃NO₂
MW: 179.64 g/mol
Melting Pt: 203 °C
MDL Number: MFCD03840364
CAS Number: 60421-23-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96bfcef95908ab87cb4e5fb2424e93f996fb5faf&vid=96bfcef95908ab87cb4e5fb2424e93f996fb5faf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR