Methyl D-3-phenyllactate ≥98.0% (by GC)

Supplier: TCI America

M2350-1G M2350-5G
TCM2350-1GEA 37.92 CAD
TCM2350-1G TCM2350-5G
Methyl D-3-phenyllactate ≥98.0% (by GC)
Methyl D-3-phenyllactate

CAS Number: 27000-00-6
MDL Number: MFCD09842268
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 115
Melting point (°C): 49
Specific rotation [a]20/D: -5 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: D-3-Phenyllactic Acid Methyl Ester, Methyl (R)-2-Hydroxy-3-phenylpropionate, (R)-2-Hydroxy-3-phenylpropionic Acid Methyl Ester

Formula: C₁₀H₁₂O₃
Boiling Pt: 115 °C
Melting Pt: 49 °C
MDL Number: MFCD09842268
CAS Number: 27000-00-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fd10037121605bface13bcfc789319df0c34ecf&vid=0fd10037121605bface13bcfc789319df0c34ecf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR