3-Formyl-6-methoxypyridine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 6-Methoxypyridine-3-carbaldehyde, 6-Methoxynicotinaldehyde, 6-Methoxypyridine-3-carboxaldehyde, 2-Methoxypyridine-5-carboxaldehyde, 6-Methoxy-3-pyridinecarboxaldehyde

M2347-1G M2347-5G
TCM2347-1GEA 85.32 CAD
TCM2347-1G TCM2347-5G
3-Formyl-6-methoxypyridine ≥98.0% (by GC)
3-Formyl-6-methoxypyridine

CAS Number: 65873-72-5
MDL Number: MFCD02683446
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 51
Flash Point (°C): 107


Other Product Names / Synonyms: 5-Formyl-2-methoxypyridine, 6-Methoxynicotinaldehyde

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 51 °C
Flash Pt: 107 °C
MDL Number: MFCD02683446
CAS Number: 65873-72-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44163cc36558aea85f2135e5a480365b24d385af&vid=44163cc36558aea85f2135e5a480365b24d385af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR