Methyl-3,4-diaminobenzoate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M2346-25G
M2346-5G
TCM2346-25GEA
314.42
CAD
TCM2346-25G
TCM2346-5G
Methyl-3,4-diaminobenzoate ≥98.0% (by GC, titration analysis)
Methyl 3,4-diaminobenzoate
CAS Number: 36692-49-6
MDL Number: MFCD00017098
Molecular Formula: C8H10N2O2
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 109
Other Product Names / Synonyms: 3,4-Diaminobenzoic Acid Methyl Ester
Formula:
C₈H₁₀N₂O₂ Melting Pt: 109 °C |
MDL Number:
MFCD00017098 CAS Number: 36692-49-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...