Methyl-6-bromopicolinate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 6-bromopicolinate, Methyl 6-bromopyridine-2-carboxylate, 6-Bromopyridine-2-carboxylic acid methyl ester, 6-Bromopicolinic acid methyl ester

M2342-1G M2342-5G
TCM2342-1GEA 30.02 CAD
TCM2342-1G TCM2342-5G
Methyl-6-bromopicolinate ≥98.0% (by GC)
Methyl 6-bromopicolinate

CAS Number: 26218-75-7
MDL Number: MFCD06203934
Molecular Formula: C7H6BrNO2
Molecular Weight: 216.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 95


Other Product Names / Synonyms: 6-Bromopyridine-2-carboxylic Acid Methyl Ester, Methyl 6-Bromopicolinate, 6-Bromopicolinic Acid Methyl Ester

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 95 °C
MDL Number: MFCD06203934
CAS Number: 26218-75-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d04fa2126fa6a76afbc31fe4b5fd75a273dc1af&vid=7d04fa2126fa6a76afbc31fe4b5fd75a273dc1af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR